Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003415.1 | NP_001003415.1 | 3 - 4 | 1164 | 78 | |
GenBank mRNA | AY648747.1 | - | 3 - 4 | 1164 | 78 | |
BC093108.1 | - | 3 - 4 | 1165 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |