Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caprin1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003484.1 | NP_001003484.1 | 3 - 4 | 331 | 90 | |
XM_005172274.4 | XP_005172331.1 | 3 - 4 | 348 | 90 | ||
XM_005172275.4 | XP_005172332.1 | 3 - 4 | 348 | 90 | ||
GenBank mRNA | BC078410.1 | - | 3 - 4 | 331 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |