Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caprin1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003484.1 | NP_001003484.1 | 7 - 8 | 886 | 112 | |
XM_005172274.4 | XP_005172331.1 | 7 - 8 | 903 | 112 | ||
XM_005172275.4 | XP_005172332.1 | 7 - 8 | 903 | 112 | ||
GenBank mRNA | BC078410.1 | - | 7 - 8 | 886 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |