Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006054.1 | NP_001006054.1 | 2 - 3 | 490 | 89 | |
GenBank mRNA | BC083278.1 | - | 2 - 3 | 490 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |