Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007151.3 | NP_001007152.1 | 1 - 2 | 195 | 87 | |
GenBank mRNA | AY640176.1 | - | 1 - 2 | 78 | 87 | |
BC108053.1 | - | 1 - 2 | 114 | 87 | ||
EB845503.1 | - | 1 - 2 | 198 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |