Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007151.3 | NP_001007152.1 | 4 - 5 | 645 | 72 | |
GenBank mRNA | AY640176.1 | - | 4 - 5 | 528 | 72 | |
BC108053.1 | - | 4 - 5 | 564 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |