Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007787.1 | NP_001007788.1 | 763 | 74 | ||
GenBank mRNA | AY572416.1 | - | 763 | 74 | ||
BC108041.1 | - | 799 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio |