Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017747.1 | NP_001017747.1 | 3 - 4 | 467 | 83 | |
GenBank mRNA | BC093204.1 | - | 3 - 4 | 467 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |