Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017801.1 | NP_001017801.1 | 3 - 4 | 488 | 78 | |
XM_005159741.1 | XP_005159798.1 | 3 - 4 | 535 | 78 | ||
GenBank mRNA | BC092880.1 | - | 3 - 4 | 488 | 78 | |
BC164311.1 | - | 2 - 3 | 381 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |