Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020480.1 | NP_001018316.1 | 1 - 2 | 119 | 125 | |
GenBank mRNA | AB194986.1 | - | 1 - 2 | 134 | 125 | |
BC056805.1 | - | 1 - 2 | 130 | 125 | ||
BC095383.1 | - | 1 - 2 | 119 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |