Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020607.1 | NP_001018443.1 | 5 - 6 | 654 | 148 | |
XM_005156332.4 | XP_005156389.1 | 3 - 4 | 698 | 148 | ||
XM_021480111.1 | XP_021335786.1 | 3 - 4 | 401 | 148 | ||
GenBank mRNA | BC095327.1 | - | 5 - 6 | 654 | 148 | |
MG957999.1 | - | 4 - 5 | 513 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |