Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020772.1 | NP_001018608.1 | 8 - 9 | 985 | 69 | |
XM_009292818.3 | XP_009291093.1 | 8 - 9 | 1225 | 69 | ||
XM_009292819.3 | XP_009291094.1 | 8 - 9 | 1084 | 69 | ||
XM_021467167.1 | XP_021322842.1 | 8 - 9 | 914 | 69 | ||
GenBank mRNA | BC095858.1 | - | 8 - 9 | 985 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |