Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020799.1 | NP_001018635.1 | 3 - 4 | 332 | 84 | |
GenBank mRNA | BC163119.1 | - | 3 - 4 | 409 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |