Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: afg1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025482.2 | NP_001020653.2 | 2 - 3 | 439 | 110 | |
GenBank mRNA | BC115066.1 | - | 2 - 3 | 439 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |