Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt7aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025540.1 | NP_001020711.1 | 1 - 2 | 71 | 90 | |
GenBank mRNA | BC163690.1 | - | 1 - 2 | 127 | 90 | |
BC163691.1 | - | 1 - 2 | 127 | 90 | ||
DQ074644.1 | - | 1 - 2 | 71 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |