Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025546.1 | NP_001020717.1 | 3 - 4 | 822 | 87 | |
XM_005160887.4 | XP_005160944.1 | 3 - 4 | 1100 | 87 | ||
GenBank mRNA | BC097244.1 | - | 3 - 4 | 822 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |