Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025546.1 | NP_001020717.1 | 6 - 7 | 1249 | 82 | |
XM_005160887.4 | XP_005160944.1 | 6 - 7 | 1527 | 82 | ||
GenBank mRNA | BC097244.1 | - | 6 - 7 | 1249 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |