Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030176.1 | NP_001025347.1 | 1 - 2 | 834 | 88 | |
XM_005160463.4 | XP_005160520.3 | 2 - 3 | 987 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |