Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emilin1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030207.1 | NP_001025378.1 | 3 - 4 | 436 | 141 | |
XM_005160756.3 | XP_005160813.1 | 3 - 4 | 776 | 141 | ||
GenBank mRNA | BC133984.1 | - | 3 - 4 | 732 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |