Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ston2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033743.1 | NP_001028915.1 | 4 - 5 | 541 | 70 | |
GenBank mRNA | BC096906.1 | - | 4 - 5 | 541 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |