Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037415.1 | NP_001032492.1 | 7 - 8 | 1899 | 115 | |
GenBank mRNA | AY928518.1 | - | 7 - 8 | 1899 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |