Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ing1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040356.1 | NP_001035446.1 | 3 - 4 | 398 | 119 | |
GenBank mRNA | BC115329.1 | - | 3 - 4 | 398 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |