Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044928.1 | NP_001038393.1 | 8 - 9 | 935 | 60 | |
XM_005159517.4 | XP_005159574.1 | 8 - 9 | 966 | 60 | ||
GenBank mRNA | BC171436.1 | - | 8 - 9 | 887 | 60 | |
FN428734.1 | - | 5 - 6 | 460 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |