Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044978.2 | NP_001038443.2 | 6 - 7 | 1405 | 78 | |
GenBank mRNA | BC134850.1 | - | 6 - 7 | 1405 | 78 | |
BC139521.1 | - | 6 - 7 | 1509 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |