Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045155.2 | NP_001038620.2 | 5 - 6 | 597 | 61 | |
XM_021475124.1 | XP_021330799.1 | 5 - 6 | 628 | 61 | ||
XM_021475125.1 | XP_021330800.1 | 2 - 3 | 202 | 61 | ||
XM_021475126.1 | XP_021330801.1 | 2 - 3 | 224 | 61 | ||
XM_021475127.1 | XP_021330802.1 | 2 - 3 | 788 | 61 | ||
XM_021475128.1 | XP_021330803.1 | 2 - 3 | 792 | 61 | ||
GenBank mRNA | BC134082.1 | - | 5 - 6 | 597 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |