Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076609.1 | NP_001070077.1 | 2 - 3 | 305 | 66 | |
XM_009298174.3 | XP_009296449.1 | 2 - 3 | 328 | 66 | ||
GenBank mRNA | BC124488.1 | - | 2 - 3 | 305 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |