Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076609.1 | NP_001070077.1 | 3 - 4 | 448 | 99 | |
XM_009298174.3 | XP_009296449.1 | 3 - 4 | 471 | 99 | ||
GenBank mRNA | BC124488.1 | - | 3 - 4 | 448 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |