Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faah2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077462.1 | NP_001070930.1 | 2 - 3 | 311 | 65 | |
GenBank mRNA | BC124392.1 | - | 2 - 3 | 311 | 65 | |
BC165762.1 | - | 2 - 3 | 296 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |