Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc29a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080572.2 | NP_001074041.2 | 6 - 7 | 894 | 71 | |
NM_001362610.1 | NP_001349539.1 | 6 - 7 | 894 | 71 | ||
GenBank mRNA | BC129376.1 | - | 6 - 7 | 894 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |