Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082855.1 | NP_001076324.1 | 5 - 6 | 1128 | 74 | |
GenBank mRNA | BC134163.1 | - | 5 - 6 | 1128 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |