Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sin3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098180.1 | NP_001091650.1 | 13 - 14 | 2257 | 63 | |
XM_005162970.4 | XP_005163027.1 | 13 - 14 | 2216 | 63 | ||
GenBank mRNA | BC139856.1 | - | 13 - 14 | 2257 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |