Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sin3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098180.1 | NP_001091650.1 | 15 - 16 | 3021 | 94 | |
XM_005162970.4 | XP_005163027.1 | 15 - 16 | 2980 | 94 | ||
GenBank mRNA | BC139856.1 | - | 15 - 16 | 3021 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |