Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sin3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098180.1 | NP_001091650.1 | 16 - 17 | 3197 | 72 | |
XM_005162970.4 | XP_005163027.1 | 16 - 17 | 3156 | 72 | ||
GenBank mRNA | BC139856.1 | - | 16 - 17 | 3197 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |