Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lctlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100014.2 | NP_001093484.1 | 4 - 5 | 465 | 106 | |
XM_005159111.3 | XP_005159168.1 | 5 - 6 | 684 | 106 | ||
XM_009293569.2 | XP_009291844.1 | 5 - 6 | 684 | 106 | ||
XM_021467671.1 | XP_021323346.1 | 5 - 6 | 684 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |