Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: upk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100063.1 | NP_001093533.1 | 2 - 3 | 270 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |