Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103111.1 | NP_001096581.1 | 5 - 6 | 1208 | 134 | |
XM_005170271.4 | XP_005170328.1 | 6 - 7 | 1293 | 134 | ||
GenBank mRNA | BC150227.1 | - | 5 - 6 | 1208 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |