Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130973.3 | NP_571048.2 | 15 - 16 | 2389 | 64 | |
GenBank mRNA | AF034606.1 | - | 15 - 16 | 2391 | 64 | |
BC162594.1 | - | 15 - 16 | 2391 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |