Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130973.3 | NP_571048.2 | 16 - 17 | 2445 | 103 | |
GenBank mRNA | AF034606.1 | - | 16 - 17 | 2447 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |