Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131032.2 | NP_571107.2 | 2 - 3 | 1179 | 107 | |
GenBank mRNA | BC091905.1 | - | 2 - 3 | 1181 | 107 | |
U89710.1 | - | 2 - 3 | 1169 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |