Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131057.1 | NP_571132.1 | 14 - 15 | 629 | 132 | |
XM_005156453.4 | XP_005156510.1 | 14 - 15 | 641 | 129 | ||
XM_017357926.2 | XP_017213415.1 | 14 - 15 | 638 | 126 | ||
XM_017357927.2 | XP_017213416.1 | 14 - 15 | 644 | 132 | ||
XM_021479102.1 | XP_021334777.1 | 13 - 14 | 605 | 132 | ||
XM_021479103.1 | XP_021334778.1 | 12 - 13 | 508 | 129 | ||
XM_021479104.1 | XP_021334779.1 | 13 - 14 | 588 | 132 | ||
XM_021479105.1 | XP_021334780.1 | 12 - 13 | 508 | 129 | ||
GenBank mRNA | AB005147.1 | - | 14 - 15 | 629 | 132 | |
BC095001.1 | - | 534 | 132 | |||
BC129275.1 | - | 13 - 14 | 596 | 132 | ||
Y12007.1 | - | 546 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |