Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131057.1 | NP_571132.1 | 8 - 9 | 338 | 115 | |
XM_005156453.4 | XP_005156510.1 | 8 - 9 | 353 | 115 | ||
XM_017357926.2 | XP_017213415.1 | 8 - 9 | 353 | 115 | ||
XM_017357927.2 | XP_017213416.1 | 8 - 9 | 353 | 115 | ||
XM_021479102.1 | XP_021334777.1 | 7 - 8 | 314 | 115 | ||
XM_021479103.1 | XP_021334778.1 | 6 - 7 | 220 | 115 | ||
XM_021479104.1 | XP_021334779.1 | 7 - 8 | 297 | 115 | ||
XM_021479105.1 | XP_021334780.1 | 6 - 7 | 220 | 115 | ||
GenBank mRNA | AB005147.1 | - | 8 - 9 | 338 | 115 | |
BC129275.1 | - | 7 - 8 | 305 | 115 | ||
Y12007.1 | - | 255 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |