Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131115.2 | NP_571190.2 | 1 - 2 | 665 | 79 | |
GenBank mRNA | BC162942.1 | - | 1 - 2 | 665 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |