Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: raf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131439.2 | NP_571514.2 | 11 - 12 | 1171 | 80 | |
XM_017353505.1 | XP_017208994.1 | 10 - 11 | 1172 | 80 | ||
XM_017353507.1 | XP_017208996.1 | 10 - 11 | 1127 | 80 | ||
XM_021469730.1 | XP_021325405.1 | 11 - 12 | 1178 | 80 | ||
GenBank mRNA | AB098700.1 | - | 11 - 12 | 1184 | 80 | |
AB098701.1 | - | 10 - 11 | 1085 | 80 | ||
AB100685.1 | - | 10 - 11 | 1086 | 80 | ||
AB116254.1 | - | 10 - 11 | 1088 | 80 | ||
AB547344.1 | - | 9 - 10 | 1045 | 80 | ||
BC054662.1 | - | 10 - 11 | 1073 | 80 | ||
X81128.1 | - | 90 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |