Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131480.1 | NP_571555.1 | 15 - 16 | 1432 | 83 | |
GenBank mRNA | AJ005692.1 | - | 15 - 16 | 1432 | 83 | |
BC060933.1 | - | 15 - 16 | 1461 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |