Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csf1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131672.1 | NP_571747.1 | 9 - 10 | 1642 | 67 | |
XM_021481056.1 | XP_021336731.1 | 10 - 11 | 1772 | 67 | ||
XM_021481057.1 | XP_021336732.1 | 10 - 11 | 1699 | 67 | ||
GenBank mRNA | AF240639.1 | - | 9 - 10 | 1642 | 67 | |
BC162803.1 | - | 9 - 10 | 1577 | 67 | ||
CU458899.1 | - | 9 - 10 | 1536 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |