Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meis1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131893.2 | NP_571968.1 | 11 - 12 | 1743 | 69 | |
GenBank mRNA | AF375871.2 | - | 11 - 12 | 1490 | 69 | |
BC060891.1 | - | 11 - 12 | 1266 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |