Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arntl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178300.1 | NP_840085.1 | 4 - 5 | 226 | 67 | |
XM_005166616.4 | XP_005166673.1 | 7 - 8 | 484 | 67 | ||
XM_009303573.3 | XP_009301848.1 | 6 - 7 | 415 | 67 | ||
XM_021477412.1 | XP_021333087.1 | 6 - 7 | 422 | 67 | ||
GenBank mRNA | AB087257.1 | - | 4 - 5 | 226 | 67 | |
BC134895.1 | - | 7 - 8 | 469 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |