Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180964.2 | NP_851295.1 | 1 | 49 | 75 | |
GenBank mRNA | AJ011789.1 | - | 1 | 49 | 75 | |
BC078260.1 | - | 1 | 75 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Both primers and probe map within a single exon |