Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrarpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181495.3 | NP_852472.1 | 1 | 356 | 127 | |
GenBank mRNA | AF509780.1 | - | 1 | 356 | 127 | |
AY187047.1 | - | 1 | 181 | 127 | ||
BC077081.1 | - | 2 | 673 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |