Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pvalb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182937.2 | NP_891982.2 | 4 - 5 | 574 | 71 | |
GenBank mRNA | AF425739.1 | - | 4 - 5 | 365 | 71 | |
BC085451.1 | - | 4 - 5 | 574 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |